Learn More
WTW™ Accessories for WTW™ MultiLine IDS/inoLab IDS
WTW offers adapter solutions for mobile energy supply, data transfer of old systems as well as for the connection of conventional pH electrodes to digital systems
Brand: WTW™ 108130
Additional Details : Weight : 0.12249kg
Description
WTW offers adapter solutions for mobile energy supply, data transfer of old systems as well as for the connection of conventional pH electrodes to digital systems. The USB adapters ADA USB/Ser and ADA USB are used to connect devices with RS 232 interface to PCs and laptops with USB interfaces, especially all lab and portable meters with RS 232 connector (portable meters 3xxi, benchtop meters inoLab™ Level 2, 730, field meters 197i, 1970i).With the ADA S7/IDS pH electrodes with S7 plug head can be connected to MultiLine™, MonoLine pH 3310 IDS as well as inoLab™ Multi 9310.Specifications
Adapter Cable | |
For connecting a SenTix™ combination electrode with S7 plug head to a MultiLine™ or inoLab™ IDS | |
digital connector | |
1.5 m |