Learn More
Digi-Sense™ Accessories for the 20250-40 Digi-Sense™ Temperature Humidity Touch-screen Recorder
Click to view available options
Product Type:
Extension Sensor Cable
Replacement Power Supply
Replacement Temperature/RH Sensor
Replacement USB Cable
Unit Size:
Each
Description
- Includes A2LA accredited NIST-Traceable Calibration Certificate
- Measures, logs, and displays temperature, humidity, and dew point
- Removable sensor for local or remote measurements using extension cable included
- Wall mount or desk mount operation
- Resident self-booting software, no installation necessary
- Comes with NIST calibration
- Stored data downloads to computer via USB cable provided in CSV format
Warranty
2 Years
Specifications
Specifications
| For Use With (Application) | Replacement AC Adapter for Temperature/RH Touch Screen Recorder with NIST Traceable Calibration |
| Includes | 5 V 2 A AC adapter |
| Product Type | Replacement Power Supply |
| For Use With (Equipment) | Temperature/RH Touch Screen Recorder with NIST Traceable Calibration (20250-40) |