Learn More
BRAND™ Accessories For Media Bottle
For use with media/ reagent bottle
765.00 NOK - 2215.00 NOK
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
15670469
|
BRAND™
122755 |
Thread Adapter |
765.00 NOK
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
15640499
|
BRAND™
122760 |
Vacuum Filter Funnel (0.2 μm) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15650499
|
BRAND™
122765 |
Vacuum Filter Funnel (0.45 μm) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Use with media bottle (122710)Specifications
Media Bottle |