Learn More
BINDER™ Access Port for BINDER™ KB ECO Series
The Access Port for BINDER KB ECO Series is an option for Series KB ECO Refrigerated Incubators with environmentally friendly Thermoelectric Cooling.
Brand: BINDER™ 8012-2116
Additional Details : Weight : 0.00100kg
Description
- Access port with silicone plug
Specifications
Access port with silicone plug - 100 mm, top | |
BINDER Chambers Series KB ECO | |
100 mm Top |