Learn More
BINDER™ Access Port for BINDER™ KB ECO Series
The Access Port for BINDER KB ECO Series is an option for Series KB ECO Refrigerated Incubators with environmentally friendly Thermoelectric Cooling.
Brand:  BINDER™ 8012-2120
5157.15 NOK valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 17532885
  • 6781.26 NOK / Each
Estimated Shipment: 03-06-2024
Description
  • Access port with silicone plug
Specifications
Specifications
Access port with silicone plug - 50 mm, left
BINDER Chambers Series KB ECO
50 mm Left