Learn More
Fisherbrand™ Clamp-ette™ Pipette Holder, Three Pipette
Product Code. 11821582 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Blue
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11821582 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11821582
  • ONLINE EXCLUSIVE PRICE
    329.00 NOK
    310.69 NOK / Pack of 1
    Save 18.31 NOK
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
Bulk Quote Request
Secures to tabletops, benches, and shelves with a twist. The Clamp-ette™ Pipette Holder is a polypropylene clamp for micropipettes.
  • Chemical-resistant polypropylene
  • Fits most major pipette brands
  • Autoclavable
Specifications
Color Blue
Holds Pipettes
Material Polypropylene
Positions 3
Stackable No
Sterility Non-sterile
Dimensions (L x W x H) 157 x 120 x 95 mm
Length (Metric) 15.7 cm
Length (English) 6.2 in.
Width (English) 4.7 in.
Height (English) 3.7 in.
Product Type Pipette Holder
Shelves None
Height (Metric) 9.5 cm
Width (Metric) 12 cm