Learn More
Testo™ 720 Temperature Meter
Description
- Port for one RTD or NTC sensor
- Wide range of optional sensors for individual requirements
- User-defined thresholds
- Audible alarm
- Optional accessories for individual adaptation like protection against aggressive media for use in laboratories
Specifications
Specifications
| Battery Life | 70 hr. |
| Battery Type | 9 V Block |
| Accuracy | ±0.2°C |
| Width (Metric) | 64 mm |