missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB4 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | ABCB4 |
---|---|
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17561356
|
Proteintech
27726-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17551356
|
Proteintech
27726-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The ABCB4 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human ABCB4. This antibody recognizes human antigen. The ABCB4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
ABCB4 | |
Polyclonal | |
Rabbit | |
5244 | |
Primary |
Western Blot, ELISA | |
Unconjugated | |
Human | |
IgG |