missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The ABCB4 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human ABCB4. This antibody recognizes human antigen. The ABCB4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
| Antigen | ABCB4 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 5244 |
| Target Species | Human |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?