Learn More
Eppendorf™ 96-well V-Bottom PCR Microplates
GREENER_CHOICE
Product Code. 10146933 Shop All Eppendorf Products
Click to view available options
Color:
Gray, White Bottom, White Bottom
White, Black Bottom, Black Bottom
White, Clear Bottom
Quantity:
240/Pk.
80/Pk.
Unit Size:
Pack of 240
Pack of 80
Product Code. 10146933
Brand: Eppendorf™ 0030601904
Only null left
Specifications
Certified Yes
Format Standard
Detection Method Quantitative Protein Analysis
Array 8 x 12
Color White, Black Bottom, Black Bottom
Material Polypropylene
No. of Wells 96
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) For PCR techniques
Max. Heat Stability 121°C
Quantity 80/Pk.