Learn More
Fisherbrand™ 96-Well Semi-Skirted PCR Plates
Product Code. 12134122 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
Yellow
Unit Size:
Pack of 25
Product Code. Color unitSize
12134122 Green Pack of 25
12144122 Yellow Pack of 25
11889261 Blue Pack of 25
12114122 Red Pack of 25
12104122 Natural Pack of 25
5 options
Product Code. 12134122
Only null left
Add to basket
Add to basket
Directly compatible with all major thermal cyclers, including ABI instruments—can be used in ABI 96-well sequencers without adapters
  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Raised-rim design
  • Sealable with adhesive films and foils, heat seals, and cap strips
  • 300μL maximum volume when used with adhesive and heat seals
  • Cut-away corner and alphanumeric grid
  • Cleanroom produced
Specifications
Format Standard
Array 8 x 12
Color Green
Product Type PCR Microplate
Skirt Style Semi
Material Polypropylene
Volume (Metric) Well 300 μL
No. of Wells 96
For Use With (Application) PCR, RT-PCR applications
Rimmed Yes
Well Color Clear