Learn More
BTX™ HT 100 Plate Handler, Manual Track Switching
Description
- Quickly optimize for better gene delivery
- Designed to work with 96 well plates
- Pulse switching technology integrated into package and gold plated contacts to mate with electroporation plate
- With an adapter to accommodate 25 well plates as well with our ECM 630 generator
- Prosess up to 96 wells in seconds
Specifications
Specifications
| Format | 25-well, 96-well |
| For Use With (Application) | High-Throughput Electroporation |
| For Use With (Equipment) | ECM630, ECM830, Gemini X2 electroporation systems |
| Product Line | BTX Electroporation |
| Product Type | Electroporation accessory |
| Voltage | 3000 VDC, 250 VAC |
| Description | HT-100 Microplate Handler |
| Display | LCD |
| Electrode Shape | Pin |
| Includes | Manual track switching, Connectors for standard pulse generators |
| Show More |