Learn More
Eppendorf™ 96-Well Flat Bottom PCR Microplates
GREENER_CHOICE
Product Code. 15294516 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Gray
White, Black
White, Clear Bottom
Quantity:
240/Pk.
80/Pk.
Unit Size:
Pack of 240
Pack of 80
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity unitSize
15294516 Gray 80/Pk. Pack of 80
15204526 White, Black 80/Pk. Pack of 80
10713224 White, Clear Bottom 80/Pk. Pack of 80
10298393 White, Clear Bottom 240/Pk. Pack of 240
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15294516 Supplier Eppendorf™ Supplier No. 0030601475
Only null left
Add to basket
Add to basket
For quantitative protein analysis
TRUSTED_SUSTAINABILITY
Specifications
Certified Yes
Format Standard
Detection Method Quantitative Protein Analysis
Array 8 x 12
Packaging 5 Bags x 16 Plates
Color Gray
Material Polypropylene
No. of Wells 96
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) For PCR techniques
Max. Heat Stability 121°C
Quantity 80/Pk.