missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well, Advanced TC Treated, Flat-Bottom Microplate

Description
Advanced TC polymer modification changes plastic surface in a way that increases primary and long-term cell adhesion and positively influences cell-dependant features and function enhanced cell attachment and higher proliferation rates facilitate rapid cell expansion even under restricted growth conditions.
- Provide conditions for improved cell adherence, accelerated cell expansion and increase overall cell yield
- For use with serum reduced and serum-free media
- Allows differentiation of semi-adherent cells
- Non-pyrogenic and non-cytotoxic
- Homogenous cell growth
- Higher transgene activity
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Culture Area | 34 mm2 |
| Array | 8 x 12 |
| Packaging | 1/Pk., 100/Cs. |
| Color | Clear |
| Material | Polystyrene |
| Volume (Metric) Well | 25 to 340 μL |
| No. of Wells | 96 |
| Sterility | Sterile |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?