Learn More
Fisherbrand™ 8mm PP Screw Seal, White, Center hole, 8-425 thread, Assembled septum
Shop All Fisherbrand Products
Click to view available options
Thickness (Metric):
1.3 mm
1.5 mm
missing translation for 'unitSize'
Pack of 1000
Specifications
Specifications
| Material (Liner) | Silicone/PTFE |
| Color | Cream, Red, White |
| Diameter (Metric) Outer | 8 mm |
| Material | Polypropylene |
| For Use With (Application) | GC, HPLC |
| Quantity | 100 |
| Thickness (Metric) | 1.5 mm |
| Type | Screw Cap |