Learn More
Fisherbrand™ 8mm PE Plug, Blue, for Micro-Insert
Product Code. 12313690 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Blue
Unit Size:
Pack of 960
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12313690 Blue Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12313690
Quantity Price
1 609.96 NOK / Pack of 960
10 560.98 NOK / Pack of 960
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with for MTRS-Micro-Insert
  • PE Plug
  • Blue
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material Polyethylene
For Use With (Equipment) MTRS-Micro-Insert
Product Type Plug