Learn More
Rephile™ 60 L Tank Level Sensor
Product Code. 15439659 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15439659 Water Purification System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15439659
  • ONLINE EXCLUSIVE PRICE
    3056.55 NOK
    2323.88 NOK / Pack of 1
    Save 732.67 NOK
    23% Off
 Restricted Item
Estimated Shipment: 21-04-2027
Add to basket
Add to basket
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
60 L Tank Level Sensor. Comparable to FTPF05381. Reuse the float.
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Water Purification System
Type Level Sensor