missing translation for 'onlineSavingsMsg'
Learn More

(6-Thien-2-ylpyridin-3-yl)methylamine dihydrochloride, 90%, Thermo Scientific™

Product Code. 10397154
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10397154

Brand: Thermo Scientific Maybridge CC54863DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 913830-32-7
Molecular Formula C10H12Cl2N2S
Molecular Weight (g/mol) 263.18
MDL Number MFCD09025878
InChI Key FWNBHEVGWIALSL-UHFFFAOYSA-N
Synonym 6-thien-2-ylpyridin-3-yl methylamine dihydrochloride, 6-thiophen-2-yl pyridin-3-yl methanamine dihydrochloride, 1-6-thiophen-2-yl pyridin-3-yl methanamine dihydrochloride, 6-thiophen-2-yl pyrid-3-ylmethylamine dihydrochloride, 5-aminomethyl-2-thien-2-yl pyridine dihydrochloride, 6-2-thienyl-3-pyridyl methylamine, chloride, chloride, 2-5-aminomethyl pyridin-2-yl thiophene dihydrochloride, 3-pyridinemethanamine,6-2-thienyl-,hydrochloride 1:2, 1-6-thiophen-2-yl pyridin-3-yl methanamine-hydrogen chloride 1/2
PubChem CID 24229545
IUPAC Name (6-thiophen-2-ylpyridin-3-yl)methanamine;dihydrochloride
SMILES C1=CSC(=C1)C2=NC=C(C=C2)CN.Cl.Cl

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 263.19
Percent Purity 90%
Chemical Name or Material (6-Thien-2-ylpyridin-3-yl)methylamine dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.