accessibility menu, dialog, popup

UserName

CAS RN 913830-32-7

SNH2NHClHCl

CAS RN 913830-32-7

IUPAC Name: 1-[6-(thiophen-2-yl)pyridin-3-yl]methanamine dihydrochloride
Synonyms: 1-[6-(thiophen-2-yl)pyridin-3-yl]methanamine dihydrochloride
Molecular Weight (g/mol): 263.18
Molecular Formula: C10H12Cl2N2S
InChi Key: FWNBHEVGWIALSL-UHFFFAOYSA-N
SMILES: Cl.Cl.NCC1=CN=C(C=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results