missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropurine 2'-deoxyriboside, 97%

Product Code. p-7042990
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
15476619 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 15476619

Brand: Thermo Scientific Alfa Aesar J64871.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4594-45-0 | C10H11ClN4O3 | 270.673 g/mol

6-Chloropurine 2'-deoxyriboside is used in the synthesis of 9-(2,3-dideoxy-2-fluoro-beta-D-threo-pentofuranosyl)adenine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloropurine 2′-deoxyriboside is used in the synthesis of 9-(2,3-dideoxy-2-fluoro-beta-D-threo-pentofuranosyl)adenine.

Notes
Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4594-45-0
Molecular Formula C10H11ClN4O3
Molecular Weight (g/mol) 270.673
MDL Number MFCD00083282
InChI Key PGEULCIODBNODW-FSDSQADBSA-N
Synonym 6-chloro-9-2'-deoxy-b-d-ribofuranosyl purine
PubChem CID 92132234
IUPAC Name (2R,3R,5R)-5-(6-chloropurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
SMILES C1C(C(OC1N2C=NC3=C2N=CN=C3Cl)CO)O

Specifications

Color White
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 270.67
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Chloropurine 2'-deoxyriboside
compliance-icons
Product Identifier
  • 6-Chloropurine 2'-deoxyriboside
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 6-Chloropurine 2-deoxyriboside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.