Learn More
MedChemExpress Evatanepag
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Specifications
Specifications
| Content And Storage | Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Quantity | 5 mg |
| CAS | 223488-57-1 |
| Molecular Weight (g/mol) | 468.57 g/mol |
| Solubility | DMSO : ≥ 32 mg/mL (68.29 mM) |
| Synonym | CP-533536 free acid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.