Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
Specifications
Specifications
| Description | S-4-104 - Includes plate buckets (x4) |
| Type | Rotor |
| Capacity | 4 x 750 mL |
| Max. RCF | 3,214 x g |
| Max. Speed | 3,900 rpm |
| For Use With (Equipment) | Centrifuge 5810 / 5810 R |
| Includes | Plate Buckets (x4) |