Learn More
GW3965 hydrochloride, MedChemExpress
Description
GW3965 hydrochloride is a potent and selective liver X receptor (LXR) agonist with EC50s of 190 nM and 30 nM for hLXRα and hLXRβ, respectively.
Specifications
Specifications
| Chemical Name or Material | GW3965 hydrochloride |
| CAS | 405911-17-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C33H32Cl2F3NO3 |
| Quantity | 50 mg |
| Solubility Information | DMSO : 100 mg/mL (161.68 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | ClC1=C(C(F)(F)F)C=CC=C1CN(CC(C2=CC=CC=C2)C3=CC=CC=C3)CCCOC4=CC(CC(O)=O)=CC=C4.[H]Cl |
| Molecular Weight (g/mol) | 618.51 |
| Formula Weight | 618.51 |
| Show More |