Learn More
Spectrum™ Labs Spectra/Por™ 3 3.5 kD MWCO Standard RC Dry Dialysis Kits
Product Code. 15370772
Change view
Click to view available options
Diameter (Metric):
11.5 mm
29 mm
34 mm
Length (Metric):
1.1 mL/cm vol./length
5 m
6.4 mL/cm vol./length
9.3 mL/cm vol./length
Quantity:
1 m Tubing/Kit
5 m Tubing/Kit
Unit Size:
1m
5m
Each
Product Code. Diameter (Metric) Length (Metric) Quantity unitSize
15370772 29 mm 6.4 mL/cm vol./length 1 m Tubing/Kit 1m
15360772 29 mm 6.4 mL/cm vol./length 5 m Tubing/Kit 5m
15380772 34 mm 9.3 mL/cm vol./length 1 m Tubing/Kit 1m
15340772 11.5 mm 1.1 mL/cm vol./length 1 m Tubing/Kit 1m
12791138 11.5 mm 1.1 mL/cm vol./length 5 m Tubing/Kit Each
11571060 34 mm 5 m 5 m Tubing/Kit 5m
6 options
Product Code. 15370772
Brand: Spectrum™ 132 724T1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
  • 1m or 5m of Spectra/Por 3 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
  • For basic dialysis applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water) or without glycerin (no soaking required)
Specifications
MWCO 3.5 kDa
Diameter (Metric) 29 mm
Length (Metric) 6.4 mL/cm vol./length
Quantity 1 m Tubing/Kit