Learn More
Spectrum™ Labs Spectra/Por™ 2 12-14 kD MWCO Standard RC Dry Dialysis Kits
Product Code. 15340762
Change view
Click to view available options
Diameter (Metric):
6.5 mm
16 mm
29 mm
67 mm
76 mm
Length (Metric):
0.32 mL/cm vol./length
2 mL/cm
34 mL/cm vol./length
46 mL/cm vol./length
5 m
6.4 mL/cm
Quantity:
1 m Tubing/Kit
5 m Tubing/Kit
Each
Unit Size:
1m
5m
Each
Product Code. Diameter (Metric) Length (Metric) Quantity unitSize
15340762 67 mm 34 mL/cm vol./length 5 m Tubing/Kit 5m
15310762 16 mm 2 mL/cm 1 m Tubing/Kit 1m
15320762 29 mm 6.4 mL/cm 1 m Tubing/Kit 1m
15390752 6.5 mm 0.32 mL/cm vol./length 5 m Tubing/Kit 5m
15300762 6.5 mm 0.32 mL/cm vol./length 1 m Tubing/Kit 1m
11541040 29 mm 5 m Each 5m
15350762 67 mm 34 mL/cm vol./length 1 m Tubing/Kit 1m
15360762 76 mm 46 mL/cm vol./length 5 m Tubing/Kit 5m
12771138 16 mm 5 m Each Each
9 options
Product Code. 15340762
Brand: Spectrum™ 132682T
Includes
  • 1m or 5m of Spectra/Por 2 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
  • For higher permeability applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water)
Specifications
MWCO 12 to 14 kDa
Diameter (Metric) 67 mm
Length (Metric) 34 mL/cm vol./length
Quantity 5 m Tubing/Kit