Learn More
Spectrum™ Labs Spectra/Por™ 1 6-8 kD MWCO Standard RC Dry Dialysis Kits
Description
Includes
- 1m or 5m of Spectra/Por 1 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
- For basic dialysis applications, or evaluating dialysis tubing for dialysis application
- Packaged dry with glycerin (soak in water)
Specifications
Specifications
| MWCO | 6 to 8 kDa |
| Diameter (Metric) | 76 mm |
| Length (Metric) | 46 mL/cm vol./length |
| Quantity | 5 m Tubing/Kit |