Learn More
Spectrum™ Labs Spectra/Por™ 7 50,000 D MWCO Standard RC Pre-Treated Dialysis Kits
Product Code. 15340712
Change view
Click to view available options
Diameter (Metric):
7.5 mm
18 mm
22 mm
Length (Metric):
0.45 mL/cm vol./length
2.5 mL/cm vol./length
3.7 mL/cm vol./length
Unit Size:
1m
Product Code. Diameter (Metric) Length (Metric) unitSize
15340712 22 mm 3.7 mL/cm vol./length 1m
15320712 7.5 mm 0.45 mL/cm vol./length 1m
15330712 18 mm 2.5 mL/cm vol./length 1m
3 options
Product Code. 15340712
Brand: Spectrum™ 132130T
Includes
  • 1meter of pre-treated tubing, a pair of closures and 5 opening picks
With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds
  • Hydrophilic, symmetric porositiy
  • Equilibrated in water
  • Packaged wet in preservative solution of 0.05% sodium azide
  • For small experiments or qualifying a membrane for a particular application
Specifications
MWCO 50 kDa
Diameter (Metric) 22 mm
Length (Metric) 3.7 mL/cm vol./length
Quantity 1 m Tubing/Kit