Learn More
Spectrum™ Labs Spectra/Por™ 2 and 4 12-14 kD MWCO Standard RC Dry Dialysis Kits
Product Code. 15330772
Change view
Click to view available options
Diameter (Metric):
20.4 mm
48 mm
Length (Metric):
18 mL/cm vol./length
3.3 mL/cm vol./length
Quantity:
1 m Tubing/Kit
5 m Tubing/Kit
Unit Size:
1m
5m
Product Code. Diameter (Metric) Length (Metric) Quantity unitSize
15330772 48 mm 18 mL/cm vol./length 1 m Tubing/Kit 1m
15320772 48 mm 18 mL/cm vol./length 5 m Tubing/Kit 5m
15300772 20.4 mm 3.3 mL/cm vol./length 1 m Tubing/Kit 1m
15390762 20.4 mm 3.3 mL/cm vol./length 5 m Tubing/Kit 5m
4 options
Product Code. 15330772
Brand: Spectrum™ 132 709T1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
  • 1m or 5m of Spectra/Por 2 or 4 dialysis tubing
  • One Standard Spectra/Por Closure (for buoyance)
  • One weighted Spectra/Por Closure (for orientation)
  • Two adhesive tubing openers
For smaller dialysis experiments or evaluating dialysis tubing for dialysis application
  • Hydrophilic, symmetric porosity
  • Ensures higher permeability and basic dialysis
  • Available as dry with glycerin (soak in water)
Specifications
MWCO 12 to 14 kDa
Diameter (Metric) 48 mm
Length (Metric) 18 mL/cm vol./length
Quantity 1 m Tubing/Kit