Learn More
Molecular Probes™ 5-IAF (5-Iodoacetamidofluorescein)
Product Code. 11554896
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. 11554896
Brand: Molecular Probes™ I30451
5-IAF (5-Iodoacetamidofluorescein)
The thiol-reactive 5-iodoacetamidofluorescein (5-IAF) can be used to produce bioconjugates with the 5-isomer of fluorescein.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 mg
Product Type 5-IAF
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Alkyl Halide, Iodoacetamide
Label or Dye FITC (Fluorescein)
Label Type Classic Dyes