Learn More
5-Fluorodeoxyuridine MP Biomedicals
Product Code. 11481922 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481922 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11481922 Supplier MP Biomedicals Supplier No. 0210555110
  • ONLINE EXCLUSIVE PRICE
    495.00 NOK
    470.63 NOK / 10mg
    Save 24.37 NOK
    4% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Specifications
CAS 50-91-9
Molecular Formula C9H11FN2O5
MDL Number MFCD00006530
Quantity 10 mg
Synonym floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd
InChI Key ODKNJVUHOIMIIZ-RRKCRQDMSA-N
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O
IUPAC Name 5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Weight (g/mol) 246.19
PubChem CID 5790
ChEBI CHEBI:60761
Formula Weight 246.2
Chemical Name or Material 5-Fluorodeoxyuridine, Inhibitor of DNA Synthesis.