missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. p-7036180
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491176 250 mg 250mg
15481176 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15491176 Supplier Thermo Scientific Alfa Aesar Supplier No. H54432.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1083168-91-5
Molecular Formula C12H17BClNO3
Molecular Weight (g/mol) 269.532
MDL Number MFCD08063054
InChI Key ZHWIZWNYOSGXSR-UHFFFAOYSA-N
Synonym 3-chloro-2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 5-chloro-6-methoxypyridine-3-boronic acid pinacol ester, 3-chloro-2-methoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-chloro-2-methoxypyridine-5-boronic acid pinacol ester, 5-chloro-6-methoxypyridin-3-yl boronic acid pinacol ester, 3-chloro-2-methoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, pyridine, 3-chloro-2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 53398545
IUPAC Name 3-chloro-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)OC)Cl

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 269.53
Percent Purity 96%
Chemical Name or Material 5-Chloro-6-methoxypyridine-3-boronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.