accessibility menu, dialog, popup

UserName

CAS RN 1083168-91-5

IUPAC Name: 3-chloro-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 3-chloro-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 269.53
Molecular Formula: C12H17BClNO3
InChi Key: ZHWIZWNYOSGXSR-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results