missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-cyano-2-hydroxy-4,6-dimethylpyridine, 98%, Thermo Scientific™

Product Code. 15437655
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15437655 250 mg 250mg
11426110 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437655 Supplier Thermo Scientific Alfa Aesar Supplier No. H27058.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23819-87-6
Molecular Formula C8H7BrN2O
Molecular Weight (g/mol) 227.06
MDL Number MFCD00052651,MFCD01117224
InChI Key DFGKQCOFARYQOA-UHFFFAOYSA-N
Synonym 5-bromo-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, 5-bromo-4,6-dimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile, 5-bromo-3-cyano-2-hydroxy-4,6-dimethylpyridine, 5-bromo-2-hydroxy-4,6-dimethylnicotinonitrile, 5-bromo-2-hydroxy-4,6-dimethyl-nicotinonitrile, 5-bromo-2-hydroxy-4,6-dimethylpyridine-3-carbonitrile, bromodimethyloxodihydropyridinecarbonitrile, 3-pyridinecarbonitrile, 5-bromo-1,2-dihydro-4,6-dimethyl-2-oxo, 5-bromo-4,6-dimethyl-2-oxohydropyridine-3-carbonitrile, acmc-20am5g
PubChem CID 737752
SMILES CC1=C(Br)C(C)=C(C#N)C(=O)N1

Specifications

Melting Point ∽256°C (decomposition)
Odor Odorless
Quantity 250 mg
IUPAC Name 5-bromo-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
Formula Weight 227.06
Percent Purity 98%
Chemical Name or Material 5-Bromo-3-cyano-2-hydroxy-4,6-dimethylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.