accessibility menu, dialog, popup

UserName

CH3BrNOHNCH3

CAS RN 23819-87-6

IUPAC Name: 5-bromo-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
Synonyms: 5-bromo-4,6-dimethyl-2-oxo-1H-pyridine-3-carbonitrile
Molecular Weight (g/mol): 227.06
Molecular Formula: C8H7BrN2O
InChi Key: DFGKQCOFARYQOA-UHFFFAOYSA-N
SMILES: CC1=C(Br)C(C)=C(C#N)C(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results