Learn More
5-Bromo-2'-deoxyuridine, Fisher BioReagents
Product Code. p-24835
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
Packaging:
Amber Glass
Unit Size:
1g
250mg
500mg
5g
Product Code. 10606005
Brand: Fisher BioReagents BP2508250
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
5-Bromo-2'-Deoxyuridine is a thymidine analog used as a mutagen in genetic research.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 59-14-3
Molecular Formula C9H11BrN2O5
Molecular Weight (g/mol) 307.10
MDL Number MFCD00006529
InChI Key WOVKYSAHUYNSMH-RRKCRQDMSA-N
Synonym 5-bromo-2'-deoxyuridine, broxuridine, bromodeoxyuridine, brdu, 5-bromodeoxyuridine, 5-brdu, budr, 5-bromouracil deoxyriboside, bromouracil deoxyriboside, 5-bromodesoxyuridine
PubChem CID 6035
ChEBI CHEBI:472552
IUPAC Name 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O
Specifications
Melting Point 191°C
Color White
Infrared Spectrum Conforms to reference
Assay Percent Range >99%
Packaging Amber Glass
Quantity 250 mg
Merck Index 15, 1461
Formula Weight 307.1
Physical Form Powder or Solid
Chemical Name or Material 5-Bromo-2'-Deoxyuridine, Off-white Powder
compliance-icons compliance-icons
Product Identifier
  • 5-Bromo-2'-deoxyuridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.