missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ 5-(and-6)-Carboxynaphthofluorescein, Succinimidyl Ester, mixed isomers

Product Code. 11580156
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 11580156

Brand: Molecular Probes™ C653

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5-(and-6)-Carboxynaphthofluorescein, Succinimidyl Ester, mixed isomers

Naphthofluorescein has emission maxima of approximately 670 nm in aqueous solution at pH 10. The fluorescence from this dye is pH dependent (pKa ∼7.6) requiring a relatively alkaline pH for maximal fluorescence. The carboxylic acid of Naphthofluorescein has emission maxima of approximately 670 nm in aqueous solution at pH 10. The fluorescence from this dye is pH dependent (pKa ∼7.6) requiring a relatively alkaline pH for maximal fluorescence. The amine reactive 5-(and-6)-carboxynaphthofluorescein succinimidyl ester can be used to couple this fluorophore to biomolecules of interest.
TRUSTED_SUSTAINABILITY

Specifications

Description 5-(and-6)-Carboxynaphthofluorescein, Succinimidyl Ester, mixed isomers, 25mg, Purple, Solid, C33H19NO9 Molecular Formula, 573.5144 Molecular Weight, Naphthalene Label, Amine Chemical Reactivity, 602/671 Excitation/Emission Wavelength, Used for Labeling Ch
Formulation Solid
For Use With (Application) Labeling chemistry, protein labeling and crosslinking
Quantity 25 mg
Product Type 5-(and-6)-Carboxynaphthofluorescein, Succinimidyl Ester, Mixed Isomers
Molecular Weight (g/mol) 573.51kDa
Content And Storage -5°C to -30°C
Label or Dye Naphthalene
Color Purple
Emission 671
Excitation 602
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.