Learn More
RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve with Square Holes, ISO Certified
Product Code. 10483992 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
4 mm
4.50 mm
4.75 mm
5 mm
5.60 mm
6.30 mm
6.70 mm
7.10 mm
8 mm
9 mm
9.50 mm
10 mm
11.2 mm
12.5 mm
13.2 mm
14 mm
16 mm
18 mm
19 mm
20 mm
22.4 mm
25 mm
26.5 mm
28 mm
31.5 mm
35.5 mm
37.5 mm
40 mm
45 mm
50 mm
53 mm
56 mm
63 mm
71 mm
75 mm
80 mm
90 mm
100 mm
106 mm
112 mm
125 mm
Unit Size:
Each
Product Code. Pore Size unitSize
10483992 45 mm Each
10700125 100 mm Each
10442272 106 mm Each
10022822 10 mm Each
10454952 11.2 mm Each
10378512 12.5 mm Each
10206612 112 mm Each
10216612 125 mm Each
10367892 13.2 mm Each
10377892 14 mm Each
10337942 16 mm Each
10246462 18 mm Each
10462842 22.4 mm Each
10472842 25 mm Each
10564593 19 mm Each
10105072 20 mm Each
10092722 26.5 mm Each
10002732 28 mm Each
10423422 35.5 mm Each
10718634 31.5 mm Each
10042962 37.5 mm Each
10565553 40 mm Each
10338132 4.50 mm Each
10388272 4.75 mm Each
10286742 4 mm Each
10758624 5.60 mm Each
10493992 50 mm Each
10398322 53 mm Each
10193622 5 mm Each
10454572 56 mm Each
10624593 6.30 mm Each
10216272 6.70 mm Each
10296412 63 mm Each
10434762 7.10 mm Each
10358372 71 mm Each
10378562 75 mm Each
10790115 80 mm Each
10318422 8 mm Each
10583823 9.50 mm Each
10154782 90 mm Each
10328422 9 mm Each
41 options
Product Code. 10483992
Brand: RETSCH 60.154.045000
Only null left
Specifications
Quantity 1Each
Pore Size 45 mm
Type Test Sieve
Certifications/Compliance ISO 3310/2
Material Stainless Steel
Height (Metric) 65mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 400 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel