Learn More
- Home
- Products
- Filters and Filtration
- Mechanical Sieves
- RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve with Square Holes, ISO Certified
RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve with Square Holes, ISO Certified
Product Code. 10206612 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
4 mm
4.50 mm
4.75 mm
5 mm
5.60 mm
6.30 mm
6.70 mm
7.10 mm
8 mm
9 mm
9.50 mm
10 mm
11.2 mm
12.5 mm
13.2 mm
14 mm
16 mm
18 mm
19 mm
20 mm
22.4 mm
25 mm
26.5 mm
28 mm
31.5 mm
35.5 mm
37.5 mm
40 mm
45 mm
50 mm
53 mm
56 mm
63 mm
71 mm
75 mm
80 mm
90 mm
100 mm
106 mm
112 mm
125 mm
Unit Size:
Each
| Product Code. | Pore Size | unitSize |
|---|---|---|
| 10206612 | 112 mm | Each |
| 10700125 | 100 mm | Each |
| 10442272 | 106 mm | Each |
| 10022822 | 10 mm | Each |
| 10454952 | 11.2 mm | Each |
| 10378512 | 12.5 mm | Each |
| 10216612 | 125 mm | Each |
| 10367892 | 13.2 mm | Each |
| 10377892 | 14 mm | Each |
| 10337942 | 16 mm | Each |
| 10246462 | 18 mm | Each |
| 10462842 | 22.4 mm | Each |
| 10472842 | 25 mm | Each |
| 10564593 | 19 mm | Each |
| 10105072 | 20 mm | Each |
| 10092722 | 26.5 mm | Each |
| 10002732 | 28 mm | Each |
| 10423422 | 35.5 mm | Each |
| 10718634 | 31.5 mm | Each |
| 10042962 | 37.5 mm | Each |
| 10565553 | 40 mm | Each |
| 10338132 | 4.50 mm | Each |
| 10388272 | 4.75 mm | Each |
| 10483992 | 45 mm | Each |
| 10286742 | 4 mm | Each |
| 10758624 | 5.60 mm | Each |
| 10493992 | 50 mm | Each |
| 10398322 | 53 mm | Each |
| 10193622 | 5 mm | Each |
| 10454572 | 56 mm | Each |
| 10624593 | 6.30 mm | Each |
| 10216272 | 6.70 mm | Each |
| 10296412 | 63 mm | Each |
| 10434762 | 7.10 mm | Each |
| 10358372 | 71 mm | Each |
| 10378562 | 75 mm | Each |
| 10790115 | 80 mm | Each |
| 10318422 | 8 mm | Each |
| 10583823 | 9.50 mm | Each |
| 10154782 | 90 mm | Each |
| 10328422 | 9 mm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Quantity | 1Each |
| Pore Size | 112 mm |
| Type | Test Sieve |
| Certifications/Compliance | ISO 3310/2 |
| Material | Stainless Steel |
| Height (Metric) | 65mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 400 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Stainless Steel |