Specifications
| Quantity | 1Each |
| Pore Size | 35 mesh, 18 mesh, 10 mesh, 4 mesh, 3/8 in., 3/4 in. |
| Type | Test Sieve |
| Certifications/Compliance | ASTM E 11 |
| Material | Stainless Steel |
| For Use With (Equipment) | Shaker AS 450 |
| Height (Metric) | 65mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 400 mm |
| Frame Material | Stainless Steel |
| Show More |