Learn More
- Home
- Products
- Filters and Filtration
- Mechanical Sieves
- RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve, ASTM Certified, Millimeter Pore Sizes
RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve, ASTM Certified, Millimeter Pore Sizes
Product Code. 10620292 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
1.18 mm
1.40 mm
1.7 mm
2 mm
2.36 mm
2.8 mm
3.35 mm
4 mm
4.75 mm
5.6 mm
6.3 mm
6.7 mm
8 mm
9.5 mm
11.20 mm
12.50 mm
13.2 mm
16 mm
19 mm
22.40 mm
25 mm
26.50 mm
31.50 mm
37.50 mm
45 mm
50 mm
53 mm
63 mm
75 mm
90 mm
100 mm
106 mm
125 mm
Unit Size:
Each
| Product Code. | Pore Size | unitSize |
|---|---|---|
| 10620292 | 90 mm | Each |
| 10024981 | 1.40 mm | Each |
| 10488341 | 1.18 mm | Each |
| 10428731 | 1.7 mm | Each |
| 10075061 | 100 mm | Each |
| 10503732 | 106 mm | Each |
| 10640462 | 125 mm | Each |
| 10249071 | 12.50 mm | Each |
| 10196311 | 11.20 mm | Each |
| 10670102 | 13.2 mm | Each |
| 10136461 | 19 mm | Each |
| 10488151 | 1 mm | Each |
| 10660452 | 16 mm | Each |
| 10583912 | 2.8 mm | Each |
| 10024991 | 2.36 mm | Each |
| 10497961 | 22.40 mm | Each |
| 10196501 | 25 mm | Each |
| 10438541 | 26.50 mm | Each |
| 10438731 | 2 mm | Each |
| 10749512 | 31.50 mm | Each |
| 10228501 | 37.50 mm | Each |
| 10468921 | 3.35 mm | Each |
| 10186071 | 4.75 mm | Each |
| 10054981 | 45 mm | Each |
| 10476041 | 4 mm | Each |
| 10390952 | 5.6 mm | Each |
| 10720283 | 50 mm | Each |
| 10730283 | 53 mm | Each |
| 10446621 | 6.3 mm | Each |
| 10770853 | 6.7 mm | Each |
| 10553342 | 63 mm | Each |
| 10640102 | 8 mm | Each |
| 10740283 | 75 mm | Each |
| 10034911 | 9.5 mm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Quantity | 1Each |
| Pore Size | 90 mm |
| Type | Test Sieve |
| Certifications/Compliance | ASTM E 11 |
| Material | Stainless Steel |
| For Use With (Equipment) | Shaker AS 450 |
| Height (Metric) | 65mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 400 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Stainless Steel |
| Show More Show Less |