Learn More
RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve, ASTM Certified, Millimeter Pore Sizes
Product Code. 10553342 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
1.18 mm
1.40 mm
1.7 mm
2 mm
2.36 mm
2.8 mm
3.35 mm
4 mm
4.75 mm
5.6 mm
6.3 mm
6.7 mm
8 mm
9.5 mm
11.20 mm
12.50 mm
13.2 mm
16 mm
19 mm
22.40 mm
25 mm
26.50 mm
31.50 mm
37.50 mm
45 mm
50 mm
53 mm
63 mm
75 mm
90 mm
100 mm
106 mm
125 mm
Unit Size:
Each
Product Code. Pore Size unitSize
10553342 63 mm Each
10024981 1.40 mm Each
10488341 1.18 mm Each
10428731 1.7 mm Each
10075061 100 mm Each
10503732 106 mm Each
10640462 125 mm Each
10249071 12.50 mm Each
10196311 11.20 mm Each
10670102 13.2 mm Each
10136461 19 mm Each
10488151 1 mm Each
10660452 16 mm Each
10583912 2.8 mm Each
10024991 2.36 mm Each
10497961 22.40 mm Each
10196501 25 mm Each
10438541 26.50 mm Each
10438731 2 mm Each
10749512 31.50 mm Each
10228501 37.50 mm Each
10468921 3.35 mm Each
10186071 4.75 mm Each
10054981 45 mm Each
10476041 4 mm Each
10390952 5.6 mm Each
10720283 50 mm Each
10730283 53 mm Each
10446621 6.3 mm Each
10770853 6.7 mm Each
10640102 8 mm Each
10740283 75 mm Each
10034911 9.5 mm Each
10620292 90 mm Each
34 options
Product Code. 10553342
Brand: RETSCH 60.167.063000
Only null left
Specifications
Quantity 1Each
Pore Size 63 mm
Type Test Sieve
Certifications/Compliance ASTM E 11
Material Stainless Steel
For Use With (Equipment) Shaker AS 450
Height (Metric) 65mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 400 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel