Learn More
RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve, ASTM Certified, Millimeter Pore Sizes
Product Code. 10488151 Shop All Retsch Products
Click to view available options
Pore Size:
1 mm
1.18 mm
1.40 mm
1.7 mm
2 mm
2.36 mm
2.8 mm
3.35 mm
4 mm
4.75 mm
5.6 mm
6.3 mm
6.7 mm
8 mm
9.5 mm
11.20 mm
12.50 mm
13.2 mm
16 mm
19 mm
22.40 mm
25 mm
26.50 mm
31.50 mm
37.50 mm
45 mm
50 mm
53 mm
63 mm
75 mm
90 mm
100 mm
106 mm
125 mm
Unit Size:
Each
Product Code. 10488151
Brand: RETSCH 60.167.001000
Only null left
Specifications
Quantity 1Each
Pore Size 1 mm
Type Test Sieve
Certifications/Compliance ASTM E 11
Material Stainless Steel
For Use With (Equipment) Shaker AS 450
Height (Metric) 65mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 400 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel