Learn More
Fisherbrand™ 4-Way Clinical Rack
Product Code. p-7161581 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Blue, Purple, Green, Red, Natural
Green
Natural
Purple
Unit Size:
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
16340402 Blue, Purple, Green, Red, Natural Pack of 5
16350402 Blue Pack of 5
16360402 Purple Pack of 5
16370402 Green Pack of 5
16380402 Natural Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16340402
The 4-Way Clinical Tube Rack is the only tube rack specifically designed to hold round, flat and conical blood collection and culture tubes. It holds multiple tube sizes without multiple racks.
  • Accommodates all brands of 13 and 16 mm round, flat and conical bottom blood collection and culture tubes.
  • Only 4-way rack available with a six way connection.
  • Link different or the same well sizes, end to end or side by side, in any orientation.
  • Customize the rack to fit the number of wells required, infinite units can be linked together or use one standalone unit.
  • Uniformly spaced wells help eliminate pipetting errors which enables more accurate results.
  • Molded alphanumeric identification for quick location and retrieval of tubes.
  • Robust, made from polypropylene that is chemical resistant and easy to clean.
  • Racks can be used in water baths and placed in freezers.
  • Assorted color pack available for color coding options.
  • Patent Pending
  • Autoclavable
Specifications
Cell Dividers No
Clean Claims Cleaned with mild detergents and autoclavable
Closure Included No
Description 4-Way Clinical Rack 5 pack
Autoclavable Autoclavable
Color Blue, Purple, Green, Red, Natural
Holds Tubes (13 mm), Tubes (16 mm)
Material Polypropylene
Positions 18
Shape Rectangular
Stackable Yes
Sterility Non-sterile
For Use With (Application) Clinical Tubes
For Use With (Equipment) Shaker
No. of Racks 5
Packaging Shrink Wrap
Quantity 5 Pk.
Temperature (Metric) Range 0°C to 121°C
Dimensions (L x W x H) 9.1 x 2.8 x 3.8 in.
Length (Metric) 23.2 cm
Length (English) 9.1 in.
Width (English) 2.8 in.
Height (English) 3.8 in.
Diameter (Metric) Well 13 mm, 17 mm
Weight (English) 0.28 lb.
Weight (Metric) 0.11 kg
Includes Rack
Product Type Blood Tube Rack
Shelves None
Height (Metric) 9.5 cm
Array Side A: 6 x 3, Side B: 6 x 2, Side C: 6 x 3, Side D: 6 x 2
Width (Metric) 7.0 cm