missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenylhydrazine, 97%, moistened with >30% water

Product Code. 10743974 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10743974 5 g 5g
10529793 25 g 25g
10290463 100 g 100g
10402903 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10743974 Supplier Thermo Scientific Acros Supplier No. 383030050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 100-16-3 | C6H7N3O2 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-16-3 , 7732-18-5
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007579
InChI Key KMVPXBDOWDXXEN-UHFFFAOYSA-N
Synonym p-nitrophenylhydrazin, p-nitrophenyl hydrazine, 4-nitrophenyl hydrazine, p-nitrophenylhydrazine, 1-4-nitrophenyl hydrazine, pnd, hydrazine, p-nitrophenyl, hydrazine, 4-nitrophenyl, p-hydrazinonitrobenzene, ccris 3139
PubChem CID 3310889
ChEBI CHEBI:66931
IUPAC Name (4-nitrophenyl)hydrazine
SMILES NNC1=CC=C(C=C1)[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown to Orange
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula NO2C6H4NHNH2
Quantity 5 g
Formula Weight 153.14
Percent Purity ≥96.0%
Physical Form Moist Lumps
Chemical Name or Material 4-Nitrophenylhydrazine, Moistened with >30% water
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Nitrophenylhydrazine, moistened with >30% water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.