accessibility menu, dialog, popup

UserName

H2NNHNOO

CAS RN 100-16-3

IUPAC Name: (4-nitrophenyl)hydrazine
Synonyms: hydrazine, (4-nitrophenyl)-
Molecular Weight (g/mol): 153.14
Molecular Formula: C6H7N3O2
InChi Key: KMVPXBDOWDXXEN-UHFFFAOYSA-N
SMILES: NNC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (4)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results