missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentyloxybenzoic acid, 98%, Thermo Scientific™

Product Code. 11497267
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497267 5 g 5g
11407277 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11497267 Supplier Thermo Scientific Alfa Aesar Supplier No. A15024.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15872-41-0
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00013990
InChI Key OZPPUPJQRJYTNY-UHFFFAOYSA-N
Synonym 4-pentyloxybenzoic acid, 4-pentyloxy benzoic acid, 4-n-pentyloxybenzoic acid, benzoic acid, 4-pentyloxy, p-pentyloxybenzoic acid, 4-n-amyloxybenzoic acid, p-pentoxybenzoic acid, p-n-pentyloxybenzoic acid, 4-amyloxybenzoic acid, p-pentyloxy benzoic acid
PubChem CID 85153
IUPAC Name 4-pentoxybenzoic acid
SMILES CCCCCOC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 123°C (N)
Quantity 5 g
Beilstein 1951836
Formula Weight 208.26
Percent Purity 98%
Chemical Name or Material 4-n-Pentyloxybenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.