accessibility menu, dialog, popup

UserName

CAS RN 15872-41-0

H3COOHO

CAS RN 15872-41-0

IUPAC Name: 4-(pentyloxy)benzoic acid
Synonyms: 4-(pentyloxy)benzoic acid
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: OZPPUPJQRJYTNY-UHFFFAOYSA-N
SMILES: CCCCCOC1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results