Learn More
4-(Methylthio)benzonitrile, 98+%, Thermo Scientific™
Product Code. 11342816
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11342816
Brand: Thermo Scientific Alfa Aesar L08979.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-(Methylthio)benzonitrile was used in the preparation of 4-cyanobenzenethiol.
Solubility
Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21382-98-9
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
MDL Number MFCD00010381
InChI Key RPVGEEHGKIFQFO-UHFFFAOYSA-N
Synonym 4-methylthio benzonitrile, 4-methylsulfanyl benzonitrile, benzonitrile, 4-methylthio, 4-methylthiobenzenecarbonitrile, 4-cyanothioanisole, 4-cyano-thioanisole, pubchem10528, 4-methylthiobenzonitrile, p-methylthio benzonitrile
PubChem CID 590960
IUPAC Name 4-methylsulfanylbenzonitrile
SMILES CSC1=CC=C(C=C1)C#N
Specifications
Melting Point 59°C to 64°C
Boiling Point 272°C
Quantity 5 g
UN Number UN3439
Beilstein 2079565
Formula Weight 149.22
Percent Purity ≥98%
Chemical Name or Material 4-(Methylthio)benzonitrile