Learn More
4-Isopropoxybenzenesulfonyl chloride, 96%
Product Code. 11369565
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11369565
Brand: Thermo Scientific Alfa Aesar L18984.03
Request Bulk or Custom Format
CAS: 98995-40-5 | C9H11ClO3S | 234.694 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 98995-40-5
Molecular Formula C9H11ClO3S
Molecular Weight (g/mol) 234.694
MDL Number MFCD03093083
InChI Key IJWCRHKAQNFJLT-UHFFFAOYSA-N
Synonym 4-isopropoxybenzenesulfonyl chloride, 4-isopropoxybenzene-1-sulfonyl chloride, 4-isopropoxyphenylsulfonyl chloride, 4-propan-2-yloxy benzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-1-methylethoxy, 4-isopropoxybenzenesulphonyl chloride, benzenesulfonyl chloride, 4-1-methylethoxy-9ci, 4-isopropoxybenzenesulfonylchloride, pubchem19078, acmc-209sbk
PubChem CID 22327297
IUPAC Name 4-propan-2-yloxybenzenesulfonyl chloride
SMILES CC(C)OC1=CC=C(C=C1)S(=O)(=O)Cl
Specifications
Melting Point 30°C to 37°C
Boiling Point 112°C to 113°C (0.01 mmHg)
Quantity 1 g
UN Number UN3261
Beilstein 3269058
Sensitivity Moisture sensitive
Formula Weight 234.7
Percent Purity 96%
Chemical Name or Material 4-Isopropoxybenzenesulfonyl chloride, 25g