CAS RN 98995-40-5
CAS RN 98995-40-5
IUPAC Name:
4-(propan-2-yloxy)benzene-1-sulfonyl chloride
Synonyms:
4-isopropoxybenzenesulfonyl chloride
Molecular Weight (g/mol):
234.69
Molecular Formula:
C9H11ClO3S
InChi Key:
IJWCRHKAQNFJLT-UHFFFAOYSA-N
SMILES:
CC(C)OC1=CC=C(C=C1)S(Cl)(=O)=O
1
–
1
of
1
results
1
4-Isopropoxybenzenesulfonyl chloride, 96%, Thermo Scientific Chemicals
CAS: 98995-40-5 Molecular Formula: C9H11ClO3S Molecular Weight (g/mol): 234.694 MDL Number: MFCD03093083 InChI Key: IJWCRHKAQNFJLT-UHFFFAOYSA-N Synonym: 4-isopropoxybenzenesulfonyl chloride,4-isopropoxybenzene-1-sulfonyl chloride,4-isopropoxyphenylsulfonyl chloride,4-propan-2-yloxy benzene-1-sulfonyl chloride,benzenesulfonyl chloride, 4-1-methylethoxy,4-isopropoxybenzenesulphonyl chloride,benzenesulfonyl chloride, 4-1-methylethoxy-9ci,4-isopropoxybenzenesulfonylchloride,pubchem19078,acmc-209sbk PubChem CID: 22327297 IUPAC Name: 4-propan-2-yloxybenzenesulfonyl chloride SMILES: CC(C)OC1=CC=C(C=C1)S(=O)(=O)Cl